Product Details

SNP ID
rs1281113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8188205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGTGCAGTGCTCCCCTTTCCC[A/G]ACCCCTCTGCAATGGCTGCTCTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH3TC1 PubMed Links

Gene Details

Gene
SH3TC1
Gene Name
SH3 domain and tetratricopeptide repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318480.1 Intron NP_001305409.1
NM_018986.4 Intron NP_061859.3
XM_006713889.2 Intron XP_006713952.1
XM_006713891.3 Intron XP_006713954.1
XM_011513488.1 Intron XP_011511790.1
XM_011513489.2 Intron XP_011511791.1
XM_017008300.1 Intron XP_016863789.1
XM_017008301.1 Intron XP_016863790.1
XM_017008302.1 Intron XP_016863791.1

View Full Product Details