Product Details

SNP ID
rs1802183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43226360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCAGCCACTCCAGGTCCTGCT[A/C]ATGGATGAGCCTGTCACCCCCAGCA
Phenotype
MIM: 607489
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CUL9 PubMed Links

Gene Details

Gene
CUL9
Gene Name
cullin 9
There are no transcripts associated with this gene.

Gene
DNPH1
Gene Name
2'-deoxynucleoside 5'-phosphate N-hydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006443.2 Intron NP_006434.1
NM_199184.1 Intron NP_954653.1

View Full Product Details