Product Details

SNP ID
rs1764344
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:225490839 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTTTTACCAACCATCCATGGGCT[A/G]TTGCTGAGTTGCGGAACTAGCCTAC
Phenotype
MIM: 609061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENAH PubMed Links

Gene Details

Gene
ENAH
Gene Name
enabled homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008493.1 8931 UTR 3 NP_001008493.1
NM_018212.4 8931 UTR 3 NP_060682.2
XM_005273187.2 8931 UTR 3 XP_005273244.1
XM_011544229.2 8931 Intron XP_011542531.2
XM_017001746.1 8931 UTR 3 XP_016857235.1
XM_017001747.1 8931 Intron XP_016857236.1
XM_017001748.1 8931 Intron XP_016857237.1
XM_017001749.1 8931 Intron XP_016857238.1
XM_017001750.1 8931 Intron XP_016857239.1
XM_017001751.1 8931 UTR 3 XP_016857240.1
XM_017001752.1 8931 Intron XP_016857241.1
XM_017001753.1 8931 UTR 3 XP_016857242.1
XM_017001754.1 8931 UTR 3 XP_016857243.1
XM_017001755.1 8931 UTR 3 XP_016857244.1
XM_017001756.1 8931 UTR 3 XP_016857245.1
XM_017001757.1 8931 UTR 3 XP_016857246.1

View Full Product Details