Product Details

SNP ID
rs1010866
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:64184393 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGGTAAGAATCTGGATTAGAGAAA[A/C]CAAAGCCATGAGCACAATCACCTTC
Phenotype
MIM: 300647
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AMER1 PubMed Links

Gene Details

Gene
AMER1
Gene Name
APC membrane recruitment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152424.3 Intron NP_689637.3

View Full Product Details