Product Details

SNP ID
rs1473760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55722467 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAATACCAGTTACTACTGCTTGCT[G/T]TCTTGGGGGAGGTTCTTTTCTTTCT
Phenotype
MIM: 300725
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RRAGB PubMed Links

Gene Details

Gene
RRAGB
Gene Name
Ras related GTP binding B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006064.4 Intron NP_006055.3
NM_016656.3 Intron NP_057740.2
XM_005261973.4 Intron XP_005262030.1
XM_017029217.1 Intron XP_016884706.1
XM_017029218.1 Intron XP_016884707.1

View Full Product Details