Product Details

SNP ID
rs883009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56164382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCTTTGGGCTGCGGCGGATCC[C/G]AGCCCCGGCCCGAGAGGCAAGGAAG
Phenotype
MIM: 609931 MIM: 609930 MIM: 601734
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYL6 PubMed Links

Gene Details

Gene
MYL6
Gene Name
myosin light chain 6
There are no transcripts associated with this gene.

Gene
MYL6B
Gene Name
myosin light chain 6B
There are no transcripts associated with this gene.

Gene
SMARCC2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005269101.1 3529 Silent Mutation CTC,CTG L1194L XP_005269158.1
XM_005269102.1 3529 Silent Mutation CTC,CTG L1193L XP_005269159.1
XM_005269103.1 3529 Silent Mutation CTC,CTG L1162L XP_005269160.1
XM_005269104.1 3529 Intron XP_005269161.1
XM_011538693.2 3529 Silent Mutation CTC,CTG L943L XP_011536995.1
XM_017019884.1 3529 Intron XP_016875373.1
XM_017019885.1 3529 Intron XP_016875374.1
XM_017019886.1 3529 Intron XP_016875375.1
XM_017019887.1 3529 Silent Mutation CTC,CTG L912L XP_016875376.1

View Full Product Details