Product Details

SNP ID
rs772701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55931222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTTGCTGTACCACCTTCACCA[C/G]CATCCATGCGACCCCAAGAGCCTTA
Phenotype
MIM: 125855
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DGKA PubMed Links

Gene Details

Gene
DGKA
Gene Name
diacylglycerol kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001345.4 147 UTR 5 NP_001336.2
NM_201444.2 147 Intron NP_958852.1
NM_201445.1 147 Intron NP_958853.1
NM_201554.1 147 Intron NP_963848.1
XM_005268688.2 147 Intron XP_005268745.1
XM_005268689.2 147 Intron XP_005268746.1
XM_005268690.2 147 Intron XP_005268747.1
XM_011537991.2 147 Intron XP_011536293.1
XM_011537993.2 147 Intron XP_011536295.1
XM_011537995.2 147 Intron XP_011536297.1
XM_017018900.1 147 Intron XP_016874389.1
XM_017018901.1 147 UTR 5 XP_016874390.1
XM_017018902.1 147 Intron XP_016874391.1
XM_017018903.1 147 Intron XP_016874392.1
XM_017018904.1 147 Intron XP_016874393.1
XM_017018905.1 147 Intron XP_016874394.1
XM_017018906.1 147 Intron XP_016874395.1
XM_017018907.1 147 Intron XP_016874396.1
XM_017018908.1 147 Intron XP_016874397.1
XM_017018909.1 147 Intron XP_016874398.1
Gene
PYM1
Gene Name
PYM homolog 1, exon junction complex associated factor
There are no transcripts associated with this gene.

View Full Product Details