Product Details

SNP ID
rs984896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56085074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCATTGCCCAACCTCCGCGTGG[G/T]GCGAGGGACCCAGGTCTACGATGGG
Phenotype
MIM: 190151
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERBB3 PubMed Links

Gene Details

Gene
ERBB3
Gene Name
erb-b2 receptor tyrosine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005915.1 507 Missense Mutation GGG,GTG G105V NP_001005915.1
NM_001982.3 507 Missense Mutation GGG,GTG G105V NP_001973.2

View Full Product Details