Product Details

SNP ID
rs1049697
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:133237922 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTTACATCGAGTAACCCCAATTCC[G/A]CCCCCACCCCAGTGCTCCTACTCCG
Phenotype
MIM: 605262 MIM: 603398
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NDRG1 PubMed Links

Gene Details

Gene
NDRG1
Gene Name
N-myc downstream regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135242.1 2224 UTR 3 NP_001128714.1
NM_001258432.1 2224 UTR 3 NP_001245361.1
NM_001258433.1 2224 UTR 3 NP_001245362.1
NM_006096.3 2224 UTR 3 NP_006087.2
XM_011516791.1 2224 UTR 3 XP_011515093.1
XM_011516792.1 2224 UTR 3 XP_011515094.1
Gene
WISP1
Gene Name
WNT1 inducible signaling pathway protein 1
There are no transcripts associated with this gene.

View Full Product Details