Product Details
- SNP ID
-
rs1045750
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:4251072 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATCTACAAGGGGAAGAAATTCAA[C/T]GCTCGGAAGGAGACGGTGCAGAACG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC94
PubMed Links
Gene Details
- Gene
- CCDC94
- Gene Name
- coiled-coil domain containing 94
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018074.4 |
204 |
Silent Mutation |
AAC,AAT |
N57N |
NP_060544.2 |
View Full Product Details