Product Details

SNP ID
rs1761802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:13109036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGTAAGTATTAGCAATCGCCA[A/G]TGGGTTTGTGGGACTCCCGGGGACC
Phenotype
MIM: 602432
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC3 PubMed Links

Gene Details

Gene
CCDC3
Gene Name
coiled-coil domain containing 3
There are no transcripts associated with this gene.

Gene
OPTN
Gene Name
optineurin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008211.1 Intron NP_001008212.1
NM_001008212.1 Intron NP_001008213.1
NM_001008213.1 Intron NP_001008214.1
NM_021980.4 Intron NP_068815.2

View Full Product Details