Product Details

SNP ID
rs1344347
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:161073048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCGGGGACCAGGAACAAGTGTT[A/G]GGCATGGGGAGAGGGAGACGGGAGA
Phenotype
MIM: 614264 MIM: 609607
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
There are no transcripts associated with this gene.

Gene
NECTIN4
Gene Name
nectin cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030916.2 Intron NP_112178.2
XM_005245508.3 Intron XP_005245565.1
XM_011510021.2 Intron XP_011508323.1
XM_011510022.2 Intron XP_011508324.1
XM_011510023.1 Intron XP_011508325.1

View Full Product Details