Product Details

SNP ID
rs1537044
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:161072867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGTGGTCAGCGTGGAGTAACTGC[A/G]GCCCTCGGGCTCTTCACTCTGGAGA
Phenotype
MIM: 614264 MIM: 609607
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
There are no transcripts associated with this gene.

Gene
NECTIN4
Gene Name
nectin cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030916.2 1522 Missense Mutation CGC,TGC R443C NP_112178.2
XM_005245508.3 1522 Missense Mutation CGC,TGC R418C XP_005245565.1
XM_011510021.2 1522 Missense Mutation CGC,TGC R444C XP_011508323.1
XM_011510022.2 1522 Missense Mutation CGC,TGC R419C XP_011508324.1
XM_011510023.1 1522 UTR 3 XP_011508325.1

View Full Product Details