Product Details

SNP ID
rs2692361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:146118552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACTCCTCATATATTAAATTTATT[A/G]TATTCACTGATTGAAGGTCCTCATA
Phenotype
MIM: 604569
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNTNAP2 PubMed Links

Gene Details

Gene
CNTNAP2
Gene Name
contactin associated protein-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014141.5 Intron NP_054860.1
XM_017011950.1 Intron XP_016867439.1

View Full Product Details