Product Details
- SNP ID
-
rs1763696
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
55
- Location
-
Chr.1:43282380 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCATCACAGTCATAGAGGATTTAGG[A/G]GAAGAGTAGCGTAGGCAGGGGGACC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf210
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74469123] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf210
- Gene Name
- chromosome 1 open reading frame 210
- Gene
- TMEM125
- Gene Name
- transmembrane protein 125
There are no transcripts associated with this gene.
View Full Product Details