Product Details

SNP ID
rs1046539
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:43363497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCCCACCCTGACCTTTGGCCCA[A/G]AAGCTACTGCTTCAGTGTGTGGGGT
Phenotype
MIM: 603618 MIM: 611813 MIM: 159530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC20 PubMed Links

Gene Details

Gene
CDC20
Gene Name
cell division cycle 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001255.2 1242 Intron NP_001246.2
Gene
ELOVL1
Gene Name
ELOVL fatty acid elongase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256399.1 1242 UTR 3 NP_001243328.1
NM_001256401.1 1242 UTR 3 NP_001243330.1
NM_001256402.1 1242 UTR 3 NP_001243331.1
NM_022821.3 1242 UTR 3 NP_073732.1
Gene
MIR6734
Gene Name
microRNA 6734
There are no transcripts associated with this gene.

Gene
MPL
Gene Name
MPL proto-oncogene, thrombopoietin receptor
There are no transcripts associated with this gene.

View Full Product Details