Product Details

SNP ID
rs4972554
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:173356289 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGTGCTGACTTGTTAAAGCTCAGC[A/C]TGTATTTCTCGGACAGCTTCCAATA
Phenotype
MIM: 609937
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDCA7 PubMed Links

Gene Details

Gene
CDCA7
Gene Name
cell division cycle associated 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031942.4 Intron NP_114148.3
NM_145810.2 Intron NP_665809.1
XM_011511957.2 Intron XP_011510259.1

View Full Product Details