Product Details
- SNP ID
-
rs1633915
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35158421 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATGGTGAGTAGTGCAGGGGCAGG[C/G]GGCGGGGACAGGACCAAGACAAACA
- Phenotype
-
MIM: 606669
MIM: 606684
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FXYD5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114719381] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FXYD5
- Gene Name
- FXYD domain containing ion transport regulator 5
- Gene
- FXYD7
- Gene Name
- FXYD domain containing ion transport regulator 7
There are no transcripts associated with this gene.
View Full Product Details