Product Details
- SNP ID
-
rs1404678
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:64342138 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATCTAAATAATCTAACTGCTCTTCT[G/A]TTGCTTTGAGGGATCTGGAAAATTT
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF736
PubMed Links
Gene Details
- Gene
- ZNF736
- Gene Name
- zinc finger protein 736
View Full Product Details