Product Details

SNP ID
rs1042106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28003792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGTACACAATTCACTCAAGCC[A/G]GCCTGAGAAGGCCTTGGATGCAGAT
Phenotype
MIM: 136351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLT3 PubMed Links

Gene Details

Gene
FLT3
Gene Name
fms related tyrosine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004119.2 3226 UTR 3 NP_004110.2
XM_011535015.2 3226 UTR 3 XP_011533317.1
XM_011535017.2 3226 UTR 3 XP_011533319.1
XM_011535018.2 3226 UTR 3 XP_011533320.1
XM_017020486.1 3226 UTR 3 XP_016875975.1
XM_017020487.1 3226 UTR 3 XP_016875976.1
XM_017020488.1 3226 UTR 3 XP_016875977.1
XM_017020489.1 3226 UTR 3 XP_016875978.1

View Full Product Details