Product Details

SNP ID
rs955225
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27986074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAAGTGCAGAACCGAGAAGCAG[A/C]CTCCACCACAGGGAGCTCCTCCCCA
Phenotype
MIM: 615804
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
URAD PubMed Links

Gene Details

Gene
URAD
Gene Name
ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5-) decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105577.1 Intron NP_001099047.1

View Full Product Details