Product Details

SNP ID
rs1131438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119310443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTTAACTTCAACTACAAGTTCG[C/G]TCTTACGAGACGGGGGTAGCGTGCT
Phenotype
MIM: 165360 MIM: 155735
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CBL PubMed Links

Gene Details

Gene
CBL
Gene Name
Cbl proto-oncogene
There are no transcripts associated with this gene.

Gene
MCAM
Gene Name
melanoma cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006500.2 1752 Missense Mutation ACC,AGC T606S NP_006491.2
XM_017017759.1 1752 Missense Mutation ACC,AGC T585S XP_016873248.1
XM_017017760.1 1752 Missense Mutation ACC,AGC T574S XP_016873249.1
XM_017017761.1 1752 Intron XP_016873250.1
XM_017017762.1 1752 Intron XP_016873251.1
Gene
MIR6756
Gene Name
microRNA 6756
There are no transcripts associated with this gene.

View Full Product Details