Product Details
- SNP ID
-
rs709933
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:232868434 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTCACCATCCATGGCAGGGTTGGC[A/G]TCCACCCGGCCTCTGCTGCCGCTTG
- Phenotype
-
MIM: 612003
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C2orf82
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73997587] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2orf82
- Gene Name
- chromosome 2 open reading frame 82
- Gene
- GIGYF2
- Gene Name
- GRB10 interacting GYF protein 2
There are no transcripts associated with this gene.
View Full Product Details