Product Details

SNP ID
rs778368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232880623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGTGATTTGAAAAGGGAAAGG[C/G]GGTCAGTGTTCAATTCTACAGGGGC
Phenotype
MIM: 605991
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C2orf82 PubMed Links

Gene Details

Gene
C2orf82
Gene Name
chromosome 2 open reading frame 82
There are no transcripts associated with this gene.

Gene
NGEF
Gene Name
neuronal guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114090.1 Intron NP_001107562.1
NM_019850.2 Intron NP_062824.2
XM_011510923.2 Intron XP_011509225.1
XM_011510924.1 Intron XP_011509226.1
XM_011510925.1 Intron XP_011509227.1

View Full Product Details