Product Details

SNP ID
rs1543992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110612052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCCAGCAAAAGTCATTATATACA[A/T]GGAAAACAGGGTAGTTCTGTGAATG
Phenotype
MIM: 614720
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK19 PubMed Links

Gene Details

Gene
CDK19
Gene Name
cyclin dependent kinase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300960.1 3755 UTR 3 NP_001287889.1
NM_001300963.1 3755 UTR 3 NP_001287892.1
NM_001300964.1 3755 UTR 3 NP_001287893.1
NM_015076.4 3755 UTR 3 NP_055891.1
XM_005266871.3 3755 UTR 3 XP_005266928.1
XM_011535630.2 3755 UTR 3 XP_011533932.1
XM_011535631.2 3755 UTR 3 XP_011533933.1
XM_011535632.1 3755 Intron XP_011533934.1
XM_017010587.1 3755 UTR 3 XP_016866076.1
XM_017010588.1 3755 UTR 3 XP_016866077.1

View Full Product Details