Product Details

SNP ID
rs1127851
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:93121676 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGATGGCCACCAAGCCCAGGCACTT[A/T]AGACCAGAGCCAGTGGCCATCCGTA
Phenotype
MIM: 602062
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CARD19 PubMed Links

Gene Details

Gene
CARD19
Gene Name
caspase recruitment domain family member 19
There are no transcripts associated with this gene.

Gene
NINJ1
Gene Name
ninjurin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004148.3 1167 UTR 3 NP_004139.2
XM_011518716.1 1167 UTR 3 XP_011517018.1

View Full Product Details