Product Details

SNP ID
rs1058700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54465265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGCGTCCAGCATGGAGGGCCCCA[A/G]ATCCAGGTGGAAGGACAGCAGCGGG
Phenotype
MIM: 609171 MIM: 616575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC42EP5 PubMed Links

Gene Details

Gene
CDC42EP5
Gene Name
CDC42 effector protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145057.3 651 Silent Mutation CTG,TTG L95L NP_659494.2
Gene
LENG8
Gene Name
leukocyte receptor cluster member 8
There are no transcripts associated with this gene.

Gene
LENG9
Gene Name
leukocyte receptor cluster member 9
There are no transcripts associated with this gene.

View Full Product Details