Product Details

SNP ID
rs1103996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109696392 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGAACTTTATCAAGTCCCCTTAT[A/G]TATTCAAAAGAAAGGAAATCAGTAT
Phenotype
MIM: 615358 MIM: 609390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AK9 PubMed Links

Gene Details

Gene
AK9
Gene Name
adenylate kinase 9
There are no transcripts associated with this gene.

Gene
FIG4
Gene Name
FIG4 phosphoinositide 5-phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014845.5 Intron NP_055660.1
XM_011536281.2 Intron XP_011534583.1
XM_017011591.1 Intron XP_016867080.1
XM_017011592.1 Intron XP_016867081.1
XM_017011593.1 Intron XP_016867082.1

View Full Product Details