Product Details

SNP ID
rs830556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109223712 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTCATGGAGTAGTAAAGCAGCAG[A/C]AAACACCACATCCAGGTTGCAGTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100996634 PubMed Links

Gene Details

Gene
LOC100996634
Gene Name
transmembrane protein FLJ37396
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277339.1 1158 Intron NP_001264268.1
XM_017010153.1 1158 UTR 5 XP_016865642.1
XM_017010154.1 1158 UTR 5 XP_016865643.1

View Full Product Details