Product Details

SNP ID
rs565030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36899110 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATATTTGCAAACTCTTCACCTG[A/C]GAAGGGACTAATATCCAGAATTAAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF345 PubMed Links

Gene Details

Gene
ZNF345
Gene Name
zinc finger protein 345
There are no transcripts associated with this gene.

Gene
ZNF829
Gene Name
zinc finger protein 829
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037232.3 Intron NP_001032309.2
NM_001171979.1 Intron NP_001165450.1
XM_005258876.4 Intron XP_005258933.1
XM_011526933.2 Intron XP_011525235.1

View Full Product Details