Product Details

SNP ID
rs1056226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41598139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCAAGGGCATAAAGAAAACTCA[A/G]ATACTAGATTTACTTCTGGGAGGGG
Phenotype
MIM: 614637 MIM: 601786
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DESI1 PubMed Links

Gene Details

Gene
DESI1
Gene Name
desumoylating isopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015704.2 3683 UTR 3 NP_056519.1
XM_005261571.4 3683 Intron XP_005261628.1
Gene
PMM1
Gene Name
phosphomannomutase 1
There are no transcripts associated with this gene.

View Full Product Details