Product Details

SNP ID
rs1291360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:13003082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGATCATCTCAGCAGTAAGCACA[C/T]TGACAGAATTGAGTTCCAGAAAGGA
Phenotype
MIM: 605826
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM234B PubMed Links

Gene Details

Gene
FAM234B
Gene Name
family with sequence similarity 234 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020853.1 Intron NP_065904.1
XM_017019706.1 Intron XP_016875195.1
Gene
HEBP1
Gene Name
heme binding protein 1
There are no transcripts associated with this gene.

Gene
HTR7P1
Gene Name
5-hydroxytryptamine receptor 7 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details