Product Details

SNP ID
rs36633
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:54142554 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCCCCTCTGAGTAGGAAGTGTGAT[C/T]TCCAGGGCTAATGCCTCCATCCCAG
Phenotype
MIM: 604910
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNOT3 PubMed Links

Gene Details

Gene
CNOT3
Gene Name
CCR4-NOT transcription complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014516.3 Intron NP_055331.1
XM_005278279.1 Intron XP_005278336.1
XM_005278280.2 Intron XP_005278337.1
XM_005278281.1 Intron XP_005278338.1
XM_005278282.2 Intron XP_005278339.1
XM_011526992.1 Intron XP_011525294.1
XM_011526993.2 Intron XP_011525295.1

View Full Product Details