Product Details

SNP ID
rs1470108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88610513 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTACATCTTCCACAAAAGAACTT[A/C]TTGGTTTTTCTCAGACCCATGGCAG
Phenotype
MIM: 610177
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AEN PubMed Links

Gene Details

Gene
AEN
Gene Name
apoptosis enhancing nuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022767.3 Intron NP_073604.3
XM_005254966.1 Intron XP_005255023.1
XM_005254967.1 Intron XP_005255024.1
XM_006720645.3 Intron XP_006720708.2
XM_011521905.2 Intron XP_011520207.1
XM_017022489.1 Intron XP_016877978.1
XM_017022490.1 Intron XP_016877979.1
Gene
LINC01586
Gene Name
long intergenic non-protein coding RNA 1586
There are no transcripts associated with this gene.

Gene
MIR1179
Gene Name
microRNA 1179
There are no transcripts associated with this gene.

Gene
MIR3529
Gene Name
microRNA 3529
There are no transcripts associated with this gene.

Gene
MIR7-2
Gene Name
microRNA 7-2
There are no transcripts associated with this gene.

View Full Product Details