Product Details

SNP ID
rs1267655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82551448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCATGGTTTGCATCAGAGTTCA[A/C]TCCTGGTGTTTCTTTTACCTTCTAT
Phenotype
MIM: 607342
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPEB1 PubMed Links

Gene Details

Gene
CPEB1
Gene Name
cytoplasmic polyadenylation element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079533.1 Intron NP_001073001.1
NM_001079534.1 Intron NP_001073002.1
NM_001079535.1 Intron NP_001073003.1
NM_001288819.1 Intron NP_001275748.1
NM_001288820.1 Intron NP_001275749.1
NM_030594.4 Intron NP_085097.3

View Full Product Details