Product Details

SNP ID
rs1010082
Assay Type
Validated
NCBI dbSNP Submissions
63
Location
Chr.1:27941358 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGCCAGGATGAAATAAGACCAGTG[T/G]ATGAAAGTCTGGGCACCTATCCAAC
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
SMPDL3B PubMed Links

Gene Details

Gene
SMPDL3B
Gene Name
sphingomyelin phosphodiesterase acid like 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009568.2 707 Intron NP_001009568.1
NM_001304579.1 707 Intron NP_001291508.1
NM_014474.3 707 Intron NP_055289.2
XM_011541259.2 707 Intron XP_011539561.1
XM_011541260.2 707 UTR 5 XP_011539562.1

View Full Product Details