Product Details

SNP ID
rs8454
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:154957128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAAAGTAGTGGAGTGAGAGAGC[A/G]AGAGGGAGCAGATCCAGCAAAAACA
Phenotype
MIM: 606903 MIM: 600560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928120 PubMed Links

Gene Details

Gene
LOC101928120
Gene Name
uncharacterized LOC101928120
There are no transcripts associated with this gene.

Gene
PBXIP1
Gene Name
PBX homeobox interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317734.1 3078 Intron NP_001304663.1
NM_001317735.1 3078 Intron NP_001304664.1
NM_020524.3 3078 Intron NP_065385.2
Gene
PYGO2
Gene Name
pygopus family PHD finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138300.3 3078 UTR 3 NP_612157.1
Gene
SHC1
Gene Name
SHC adaptor protein 1
There are no transcripts associated with this gene.

View Full Product Details