Product Details

SNP ID
rs1619016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7647235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGAGGCATGGATGATCACCATA[C/T]CTTACCAGATCTTTTCTTAGTCATG
Phenotype
MIM: 182331
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP1B2 PubMed Links

Gene Details

Gene
ATP1B2
Gene Name
ATPase Na+/K+ transporting subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303263.1 Intron NP_001290192.1
NM_001678.4 Intron NP_001669.3

View Full Product Details