Product Details

SNP ID
rs1042526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7669572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCAGGGGAGGGAGAGATGGGGG[A/T]GGGAGGCTGTCAGTGGGGAACAAGA
Phenotype
MIM: 191170
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TP53 PubMed Links

Gene Details

Gene
TP53
Gene Name
tumor protein p53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000546.5 1161 UTR 3 NP_000537.3
NM_001126112.2 1161 UTR 3 NP_001119584.1
NM_001126113.2 1161 UTR 3 NP_001119585.1
NM_001126114.2 1161 UTR 3 NP_001119586.1
NM_001126115.1 1161 UTR 3 NP_001119587.1
NM_001126116.1 1161 UTR 3 NP_001119588.1
NM_001126117.1 1161 UTR 3 NP_001119589.1
NM_001126118.1 1161 UTR 3 NP_001119590.1
NM_001276695.1 1161 UTR 3 NP_001263624.1
NM_001276696.1 1161 UTR 3 NP_001263625.1
NM_001276697.1 1161 UTR 3 NP_001263626.1
NM_001276698.1 1161 UTR 3 NP_001263627.1
NM_001276699.1 1161 UTR 3 NP_001263628.1
NM_001276760.1 1161 UTR 3 NP_001263689.1
NM_001276761.1 1161 UTR 3 NP_001263690.1

View Full Product Details