Product Details

SNP ID
rs1060231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4771977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTCTCCCTCATTACCCTCTGC[C/T]TCGTCTGCATTGTGGCCAACGCCCT
Phenotype
MIM: 604657
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TM4SF5 PubMed Links

Gene Details

Gene
TM4SF5
Gene Name
transmembrane 4 L six family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003963.2 86 Missense Mutation CTC,TTC L19F NP_003954.2

View Full Product Details