Product Details

SNP ID
rs1170443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68539392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATCAGAATTGGAGATAACCGAG[C/G]CTTCGGCGGGGGCGGGAGGAGCTGC
Phenotype
MIM: 609451
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZFP90 PubMed Links

Gene Details

Gene
ZFP90
Gene Name
ZFP90 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305203.1 180 UTR 5 NP_001292132.1
NM_001305204.1 180 UTR 5 NP_001292133.1
NM_001305206.1 180 UTR 5 NP_001292135.1
NM_001305207.1 180 UTR 5 NP_001292136.1
NM_001305208.1 180 UTR 5 NP_001292137.1
NM_133458.3 180 UTR 5 NP_597715.2
XM_005255804.2 180 UTR 5 XP_005255861.1
XM_017022952.1 180 Intron XP_016878441.1

View Full Product Details