Product Details

SNP ID
rs1056767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62139835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAACACATACACCAGCTCCTTGAA[A/G]GCACCCTGCCGCGCCCAGCCAAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLRMTP1 PubMed Links

Gene Details

Gene
POLRMTP1
Gene Name
RNA polymerase mitochondrial pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details