Product Details

SNP ID
rs1800902
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:58840400 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGGCAGACCTTGAGCTGTCCCTCCC[C/T]GAGTGCCTAGAGTACGAGGAAGAGT
Phenotype
MIM: 139320 MIM: 610540
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAS PubMed Links

Gene Details

Gene
GNAS
Gene Name
GNAS complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000516.5 720 Intron NP_000507.1
NM_001077488.3 720 Intron NP_001070956.1
NM_001077489.3 720 Intron NP_001070957.1
NM_001077490.2 720 Intron NP_001070958.1
NM_001309840.1 720 Intron NP_001296769.1
NM_001309842.1 720 Intron NP_001296771.1
NM_001309861.1 720 Intron NP_001296790.1
NM_001309883.1 720 Intron NP_001296812.1
NM_016592.3 720 Silent Mutation CCC,CCT P98P NP_057676.1
NM_080425.3 720 Intron NP_536350.2
NM_080426.3 720 Intron NP_536351.1
XM_017027812.1 720 Intron XP_016883301.1
XM_017027813.1 720 Intron XP_016883302.1
XM_017027814.1 720 Intron XP_016883303.1
XM_017027815.1 720 Intron XP_016883304.1
XM_017027816.1 720 Intron XP_016883305.1
XM_017027817.1 720 Intron XP_016883306.1
XM_017027818.1 720 Intron XP_016883307.1
XM_017027819.1 720 Intron XP_016883308.1
XM_017027820.1 720 Intron XP_016883309.1
XM_017027821.1 720 Silent Mutation CCC,CCT P98P XP_016883310.1
XM_017027822.1 720 Silent Mutation CCC,CCT P98P XP_016883311.1
Gene
GNAS-AS1
Gene Name
GNAS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details