Product Details

SNP ID
rs13151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101281950 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGGAGGCTGATGGTGGACTGCT[C/G]TCATCACTTCCGTCATCATCTGGTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF149 PubMed Links

Gene Details

Gene
RNF149
Gene Name
ring finger protein 149
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173647.3 1196 Missense Mutation GAC,GAG D356E NP_775918.2
XM_005263920.2 1196 Missense Mutation GAC,GAG D356E XP_005263977.1
XM_005263921.3 1196 Missense Mutation GAC,GAG D356E XP_005263978.1
XM_011510990.2 1196 Missense Mutation GAC,GAG D195E XP_011509292.1
Gene
SNORD89
Gene Name
small nucleolar RNA, C/D box 89
There are no transcripts associated with this gene.

View Full Product Details