Product Details

SNP ID
rs972446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55569581 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCTGAAGCCAATGACTATCAAAA[A/C]CTCTATGGATGTATATGTGTGCAAT
Phenotype
MIM: 611676
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PDCL2 PubMed Links

Gene Details

Gene
PDCL2
Gene Name
phosducin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152401.2 Intron NP_689614.2
XM_005265728.3 Intron XP_005265785.1
XM_011534374.2 Intron XP_011532676.1
XM_017007745.1 Intron XP_016863234.1

View Full Product Details