Product Details
- SNP ID
-
rs832099
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:97991868 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGTAAAAAAAAACTTAAACCAAT[A/G]TGAATAGATTTCATTGAAGTTATGG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GABRR3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11411389] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GABRR3
- Gene Name
- gamma-aminobutyric acid type A receptor rho3 subunit (gene/pseudogene)
View Full Product Details