Product Details

SNP ID
rs906180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81181316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGACATTGAGCTGCTGCCCAC[A/G]GACATGAGACACCACGGTACAGCAT
Phenotype
MIM: 613479
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AATK-AS1 PubMed Links

Gene Details

Gene
AATK-AS1
Gene Name
AATK antisense RNA 1
There are no transcripts associated with this gene.

Gene
CEP131
Gene Name
centrosomal protein 131
There are no transcripts associated with this gene.

View Full Product Details