Product Details

SNP ID
rs820274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:81496552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTCCTTCCTCCTTTTAGTCTGC[A/G]AGTATGGGAGGTATGAGTGATCTCA
Phenotype
MIM: 607839
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBE1 PubMed Links

Gene Details

Gene
GBE1
Gene Name
glucan (1,4-alpha-), branching enzyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000158.3 Intron NP_000149.3

View Full Product Details