Product Details

SNP ID
rs1134043
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:49162704 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTCACCAGGGCCCTAGAGAGGCACC[G/T]GGAGTCCTCAAGATTGTGGAGTCCA
Phenotype
MIM: 613169
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC71 PubMed Links

Gene Details

Gene
CCDC71
Gene Name
coiled-coil domain containing 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022903.3 1643 UTR 3 NP_075054.3
Gene
KLHDC8B
Gene Name
kelch domain containing 8B
There are no transcripts associated with this gene.

Gene
LAMB2P1
Gene Name
laminin subunit beta 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details