Product Details

SNP ID
rs906148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82617082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGGCCAGTAGGAGAGTATTTAT[C/T]TCCCCTCACTGGCCCTAATGATTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00575 PubMed Links

Gene Details

Gene
LINC00575
Gene Name
long intergenic non-protein coding RNA 575
There are no transcripts associated with this gene.

View Full Product Details